Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-53
Entrez Gene ID
191952
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_Y49C4A.6
Chromosome Location
Chr.V: 693541 - 694725 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070896.3 NP_503297.3 1 - 2 196 127

View Full Product Details