Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-58
Entrez Gene ID
187746
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_R09E12.2
Chromosome Location
Chr.V: 774995 - 777272 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070922.1 NP_503323.1 2 - 3 537 178

View Full Product Details