Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-38
Entrez Gene ID
191945
Gene Name
Serpentine receptor class J-38
Gene Aliases
CELE_T02B11.5
Chromosome Location
Chr.V: 882176 - 884424 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070948.1 NP_503349.1 1 - 2 190 67

View Full Product Details