Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-32
Entrez Gene ID
178595
Gene Name
Zinc metalloproteinase nas-32
Gene Aliases
CELE_T02B11.7
Chromosome Location
Chr.V: 894909 - 899187 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070950.5 NP_503351.3 1 - 2 124 136

View Full Product Details