Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ergo-1
Entrez Gene ID
178602
Gene Name
protein-coding
Gene Aliases
CELE_R09A1.1
Chromosome Location
Chr.V: 1007317 - 1018358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313235.3 NP_001300164.1 3 - 4 1177 72
NM_001313236.3 NP_001300165.1 2 - 3 334 72
NM_070961.9 NP_503362.2 6 - 7 2594 72

View Full Product Details