Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-22
Entrez Gene ID
188704
Gene Name
protein-coding
Gene Aliases
CELE_T21H3.4
Chromosome Location
Chr.V: 1141733 - 1146501 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070984.5 NP_503385.3 6 - 7 978 78

View Full Product Details