Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cmd-1
Entrez Gene ID
178614
Gene Name
protein-coding
Gene Aliases
CELE_T21H3.3
Chromosome Location
Chr.V: 1156271 - 1158189 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070985.7 NP_503386.1 3 - 4 459 76
GenBank mRNA AJ132193.1 - 3 - 4 481 76

View Full Product Details