Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
egl-2
Entrez Gene ID
178624
Gene Name
protein-coding
Gene Aliases
CELE_F16B3.1
Chromosome Location
Chr.V: 1289373 - 1299332 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380597.1 NP_001368562.1 10 - 11 1324 76
NM_071001.5 NP_503402.3 10 - 11 1439 76

View Full Product Details