Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-3
Entrez Gene ID
190698
Gene Name
SyNapTotagmin
Gene Aliases
CELE_Y75B7AL.3
Chromosome Location
Chr.V: 1358178 - 1371014 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383276.1 3 - 4 351 131

View Full Product Details