Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-5
Entrez Gene ID
178631
Gene Name
protein-coding
Gene Aliases
CELE_R12A1.2
Chromosome Location
Chr.V: 1393112 - 1411559 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313245.4 NP_001300174.1 4 - 5 554 141
NM_001313246.3 NP_001300175.1 4 - 5 464 141
NM_001313247.3 NP_001300176.1 6 - 7 659 141
NM_071008.5 NP_503409.1 6 - 7 755 141

View Full Product Details