Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ges-1
Entrez Gene ID
178633
Gene Name
Gut esterase 1
Gene Aliases
CELE_R12A1.4
Chromosome Location
Chr.V: 1427649 - 1432137 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071010.7 NP_503411.1 2 - 3 452 85

View Full Product Details