Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- unc-60
- Entrez Gene ID
-
178640
- Gene Name
- Actin-depolymerizing factor 2, isoform c
- Gene Aliases
-
CELE_C38C3.5
- Chromosome Location
-
Chr.V: 1475086 - 1479587 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071026.10 |
NP_503427.2 |
2 - 3 |
138 |
72 |
- Gene Symbol
- F53E2.2
- Entrez Gene ID
-
41657006
- Gene Name
- Actin-depolymerizing factor 1, isoforms a/b
- Gene Aliases
-
CELE_F53E2.2
- Chromosome Location
-
Chr.V: 1475086 - 1479577 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071024.8 |
NP_503425.2 |
5 - 6 |
1442 |
72 |
View Full Product Details