Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-91
Entrez Gene ID
186991
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K04F1.4
Chromosome Location
Chr.V: 1679208 - 1680870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071079.4 NP_503480.3 2 - 3 296 149

View Full Product Details