Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-25
Entrez Gene ID
188156
Gene Name
protein-coding
Gene Aliases
CELE_T06A1.4
Chromosome Location
Chr.V: 1938263 - 1953748 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071134.5 NP_503535.2 2 - 3 205 81
GenBank mRNA EF471978.1 - 1 - 2 101 81

View Full Product Details