Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-20
Entrez Gene ID
183492
Gene Name
protein-coding
Gene Aliases
CELE_C45H4.11
Chromosome Location
Chr.V: 2147095 - 2148626 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313133.3 NP_001300062.1 3 - 4 516 111
NM_001313134.3 NP_001300063.1 1 - 2 102 111
NM_071179.4 NP_503580.2 4 - 5 624 111
NR_132529.1 3 - 4 354 111

View Full Product Details