Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-19
Entrez Gene ID
183496
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C45H4.16
Chromosome Location
Chr.V: 2164788 - 2166779 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071187.2 NP_503588.1 2 - 3 336 85

View Full Product Details