Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33C2
Entrez Gene ID
178695
Gene Name
protein-coding
Gene Aliases
CELE_C45H4.17
Chromosome Location
Chr.V: 2181366 - 2186599 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071193.3 NP_503594.1 4 - 5 1856 101

View Full Product Details