Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp-33C12
Entrez Gene ID
189364
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y5H2B.6
Chromosome Location
Chr.V: 2201435 - 2204400 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071198.3 NP_503599.1 1 - 2 221 97

View Full Product Details