Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-40
Entrez Gene ID
189365
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y5H2B.7
Chromosome Location
Chr.V: 2205030 - 2206281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071199.2 NP_503600.2 3 - 4 448 134

View Full Product Details