Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33C3
Entrez Gene ID
3565571
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F41B5.4
Chromosome Location
Chr.V: 2242778 - 2244865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071214.5 NP_503615.1 3 - 4 1410 129

View Full Product Details