Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-290
Entrez Gene ID
183034
Gene Name
Nuclear receptor domain-containing protein
Gene Aliases
CELE_C29G2.5
Chromosome Location
Chr.V: 2574181 - 2575410 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071304.3 NP_503705.3 4 - 5 454 114

View Full Product Details