Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-4
Entrez Gene ID
183033
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_C29G2.4
Chromosome Location
Chr.V: 2577903 - 2579816 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071305.2 NP_503706.2 1 - 2 145 112

View Full Product Details