Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-27
Entrez Gene ID
182840
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_C24B9.6
Chromosome Location
Chr.V: 2719696 - 2720863 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071337.1 NP_503738.1 2 - 3 152 105

View Full Product Details