Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-61
Entrez Gene ID
182843
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_C24B9.11
Chromosome Location
Chr.V: 2738721 - 2740328 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071343.4 NP_503744.3 1 - 2 235 129

View Full Product Details