Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-26
Entrez Gene ID
187605
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_R05D8.4
Chromosome Location
Chr.V: 2751256 - 2753107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071346.2 NP_503747.2 2 - 3 198 96

View Full Product Details