Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-28
Entrez Gene ID
187606
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_R05D8.5
Chromosome Location
Chr.V: 2755701 - 2757276 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071348.3 NP_503749.3 3 - 4 349 183

View Full Product Details