Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-58
Entrez Gene ID
187607
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_R05D8.6
Chromosome Location
Chr.V: 2758174 - 2759556 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071349.1 NP_503750.1 3 - 4 671 84

View Full Product Details