Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-45
Entrez Gene ID
191949
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_T03D3.6
Chromosome Location
Chr.V: 2835312 - 2836601 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071367.2 NP_503768.1 1 - 2 185 133

View Full Product Details