Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-53
Entrez Gene ID
178744
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_T03D3.1
Chromosome Location
Chr.V: 2849343 - 2852028 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071370.5 NP_503771.1 6 - 7 1089 101

View Full Product Details