Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
folt-2
Entrez Gene ID
178745
Gene Name
Folate-like transporter 2
Gene Aliases
CELE_F37B4.7
Chromosome Location
Chr.V: 2872465 - 2875600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071379.6 NP_503780.1 4 - 5 1294 82

View Full Product Details