Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
str-39
Entrez Gene ID
191975
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F37B4.12
Chromosome Location
Chr.V: 2877804 - 2879901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071381.1 NP_503782.1 1 - 2 481 96

View Full Product Details