Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srab-7
Entrez Gene ID
183267
Gene Name
protein-coding
Gene Aliases
CELE_C36C5.11
Chromosome Location
Chr.V: 3153485 - 3155172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071466.1 NP_503867.1 2 - 3 450 113

View Full Product Details