Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-63
Entrez Gene ID
178761
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_T28A11.15
Chromosome Location
Chr.V: 3254602 - 3256151 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071493.4 NP_503894.2 2 - 3 275 100

View Full Product Details