Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-3
Entrez Gene ID
185309
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F35F10.2
Chromosome Location
Chr.V: 3308469 - 3309689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071514.2 NP_503915.2 2 - 3 339 115

View Full Product Details