Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-2
Entrez Gene ID
3565938
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_C02A12.8
Chromosome Location
Chr.V: 3471870 - 3475121 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322522.2 NP_001309470.1 3 - 4 370 84

View Full Product Details