Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-31
Entrez Gene ID
3565628
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C02A12.9
Chromosome Location
Chr.V: 3476405 - 3477596 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026190.1 NP_001021361.1 3 - 4 537 140

View Full Product Details