Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lys-7
Entrez Gene ID
178772
Gene Name
Lysozyme-like protein 7
Gene Aliases
CELE_C02A12.4
Chromosome Location
Chr.V: 3481428 - 3482726 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071571.8 NP_503972.1 2 - 3 324 66

View Full Product Details