Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-32
Entrez Gene ID
182097
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C02A12.5
Chromosome Location
Chr.V: 3485549 - 3486957 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071572.5 NP_503973.2 3 - 4 586 106

View Full Product Details