Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-66
Entrez Gene ID
188824
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_T24A6.5
Chromosome Location
Chr.V: 3530703 - 3532380 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071585.1 NP_503986.1 2 - 3 632 107

View Full Product Details