Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-21
Entrez Gene ID
191901
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_T24A6.4
Chromosome Location
Chr.V: 3549853 - 3551306 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071593.5 NP_503994.1 3 - 4 770 89

View Full Product Details