Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-41
Entrez Gene ID
186592
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_F59B1.7
Chromosome Location
Chr.V: 3634559 - 3637186 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071620.3 NP_504021.1 2 - 3 262 80

View Full Product Details