Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-23
Entrez Gene ID
191606
Gene Name
protein-coding
Gene Aliases
CELE_F59B1.9
Chromosome Location
Chr.V: 3651656 - 3659572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330798.2 NP_001317822.1 1 - 2 204 74
NM_071623.5 NP_504024.2 1 - 2 271 74
GenBank mRNA AY519853.1 - 1 - 2 207 74

View Full Product Details