Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-55
Entrez Gene ID
178792
Gene Name
protein-coding
Gene Aliases
CELE_Y39H10A.2
Chromosome Location
Chr.V: 3761139 - 3766315 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330807.1 NP_001317839.1 9 - 10 1588 67
NM_001330808.1 NP_001317840.1 2 - 3 613 67
NM_071640.2 NP_504041.1 9 - 10 1582 67

View Full Product Details