Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-7
Entrez Gene ID
187706
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_R08F11.5
Chromosome Location
Chr.V: 3806261 - 3807752 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071652.1 NP_504053.1 1 - 2 615 94

View Full Product Details