Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-6
Entrez Gene ID
186715
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H10D18.3
Chromosome Location
Chr.V: 3808636 - 3810708 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071653.1 NP_504054.1 2 - 3 812 144

View Full Product Details