Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-63
Entrez Gene ID
187094
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_K07C6.6
Chromosome Location
Chr.V: 3931626 - 3933394 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071689.2 NP_504090.2 1 - 2 203 127

View Full Product Details