Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35B3
Entrez Gene ID
187092
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K07C6.2
Chromosome Location
Chr.V: 3945353 - 3947044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071694.1 NP_504095.1 1 - 2 176 68

View Full Product Details