Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dach-1
Entrez Gene ID
188350
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T09H2.1
Chromosome Location
Chr.V: 3950023 - 3952097 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071696.3 NP_504097.1 5 - 6 928 123

View Full Product Details