Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A7
Entrez Gene ID
181849
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0213.12
Chromosome Location
Chr.V: 3961681 - 3963815 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071700.3 NP_504101.1 5 - 6 1094 88

View Full Product Details