Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A8
Entrez Gene ID
181850
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0213.14
Chromosome Location
Chr.V: 3964797 - 3967880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071701.5 NP_504102.2 1 - 2 155 115

View Full Product Details